Learn More
Cayman Chemical Phosphoramidon (sodium salt), Purity: ≥98%, CAS: 164204-38-0, 10 mg
SDP
Catalog No. 502969611
Encompass_Preferred
A natural and potent inhibitor of NEP and NEP2, with Ki values of 2 nM for both enzymes; also a weak inhibitor of the related zinc metalloprotease ECE (Ki = 3.5 µM); has little or no effect on a variety of other proteases
Catalog No. 50-296-9611 Supplier Cayman Chemical Supplier No. 1511310 MG
A natural and potent inhibitor of NEP and NEP2, with Ki values of 2 nM for both enzymes; also a weak inhibitor of the related zinc metalloprotease ECE (Ki = 3.5 µM); has little or no effect on a variety of other proteases