Learn More
Medchemexpress LLC Phosphoramidous acid, N,N-bis(1-methylethyl)-, 2-cyanoethyl 6-[[(4-methoxyphenyl)diphenylmethyl]amino]hexyl ester | 114616-27-2 | 98.0% | 589.75 | 250 MG
SDP
Catalog No. 5000402907
Encompass_Preferred
MMT-Hexylaminolinker Phosphoramidite is a cleavable linker that can be used to synthesize oligonucleotide gapmers or other nucleic acid molecules. It is for research use only.
  • Cleavable linker.
  • Can be used to synthesize oligonucleotide gapmers or other nucleic acid molecules.
  • Purity: 98.0%.
  • Available in solid or liquid form.
Catalog No. 50-004-02907 Supplier Medchemexpress LLC Supplier No. 250MGHY21242
MMT-Hexylaminolinker Phosphoramidite is a cleavable linker that can be used to synthesize oligonucleotide gapmers or other nucleic acid molecules. It is for research use only.
  • Cleavable linker.
  • Can be used to synthesize oligonucleotide gapmers or other nucleic acid molecules.
  • Purity: 98.0%.
  • Available in solid or liquid form.