Learn More
Medchemexpress LLC Phosphoramidous acid, N,N-bis(1-methylethyl)-, bis(phenylmethyl) ester | 108549-23-1 | ≥98.0% | 345.42 | 25 G
SDP
Catalog No. 5000451471
Encompass_Preferred
Dibenzyl diisopropylphosphoramidite is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It may be utilized for the preparation of phosphopeptides and is also employed in the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. Additionally, it is useful for nucleotide coupling.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • May be used for the preparation of phosphopeptides
  • Used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1
  • Useful for nucleotide coupling
Catalog No. 50-004-51471 Supplier Medchemexpress LLC Supplier No. 25GHYW013777
Dibenzyl diisopropylphosphoramidite is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It may be utilized for the preparation of phosphopeptides and is also employed in the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. Additionally, it is useful for nucleotide coupling.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • May be used for the preparation of phosphopeptides
  • Used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1
  • Useful for nucleotide coupling