Learn More
Medchemexpress LLC Phosphoribosyl pyrophosphate pentasodium | 108321-05-7 | 99.4% | 499.98 | 5 MG
SDP
Catalog No. 5000367010
Encompass_Preferred
Phosphoribosyl pyrophosphate (PRPP) pentasodium is an important metabolite required in the biosynthesis of purine and pyrimidine nucleotides, the amino acids histidine and tryptophan, and the cofactors NAD and NADP. It is an important intermediate in cellular metabolism, synthesized by PRPP synthase (ribose 5-phosphate + ATP → PRPP + AMP). PRPP is ubiquitously found in living organisms and is used in substitution reactions with the formation of glycosidic bonds. It is also utilized in the biosynthesis of tetrahydromethanopterin, arabinosyl monophosphodecaprenol, and certain aminoglycoside antibiotics.
  • Important metabolite in the biosynthesis of purine and pyrimidine nucleotides, histidine, tryptophan, NAD, and NADP.
  • Intermediate in cellular metabolism.
  • For research use only.
Catalog No. 50-003-67010 Supplier Medchemexpress LLC Supplier No. HYW0130465MG
Phosphoribosyl pyrophosphate (PRPP) pentasodium is an important metabolite required in the biosynthesis of purine and pyrimidine nucleotides, the amino acids histidine and tryptophan, and the cofactors NAD and NADP. It is an important intermediate in cellular metabolism, synthesized by PRPP synthase (ribose 5-phosphate + ATP → PRPP + AMP). PRPP is ubiquitously found in living organisms and is used in substitution reactions with the formation of glycosidic bonds. It is also utilized in the biosynthesis of tetrahydromethanopterin, arabinosyl monophosphodecaprenol, and certain aminoglycoside antibiotics.
  • Important metabolite in the biosynthesis of purine and pyrimidine nucleotides, histidine, tryptophan, NAD, and NADP.
  • Intermediate in cellular metabolism.
  • For research use only.