Learn More
Hach Company Phosphoric Acid, 10%, 100 mL MDB
Supplier:  Hach Company 1476932
Encompass
For phenol distillation and wastewater sample preservation. 100 mL Marked Dropping Bottle
Catalog No. 50-199-5112