Learn More
Medchemexpress LLC Phosphoric acid, 2-ethylhexyl diphenyl ester | 1241-94-7 | 94.3% | 362.41 | 100 MG
SDP
Catalog No. 5000453500
Encompass_Preferred
2-Ethylhexyl diphenyl phosphate is an organophosphate flame retardant (OPFR) and a PPARG agonist. It also inhibits ERRγ transcriptional activity. This compound upregulates 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion, making it suitable for studies of female reproduction and fetal development.
  • Organophosphate flame retardant
  • PPARG agonist
  • Inhibits ERRγ transcriptional activity
  • Upregulates 3β-HSD1
  • Upregulates human chorionic gonadotropin (hCG)
  • Upregulates progesterone secretion
  • Suitable for studies of female reproduction and fetal development
Catalog No. 50-004-53500 Supplier Medchemexpress LLC Supplier No. HYW009066100MG
2-Ethylhexyl diphenyl phosphate is an organophosphate flame retardant (OPFR) and a PPARG agonist. It also inhibits ERRγ transcriptional activity. This compound upregulates 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion, making it suitable for studies of female reproduction and fetal development.
  • Organophosphate flame retardant
  • PPARG agonist
  • Inhibits ERRγ transcriptional activity
  • Upregulates 3β-HSD1
  • Upregulates human chorionic gonadotropin (hCG)
  • Upregulates progesterone secretion
  • Suitable for studies of female reproduction and fetal development