Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphoric acid 2-hydroxyethyl methacrylate ester

Catalog No. 695890250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-589-0250ML 250 mL
69-589-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-589-0250ML Supplier Sigma Aldrich Supplier No. 695890250ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphoric acid 2-hydroxyethyl methacrylate ester is a phosphoric acid based ester which is composed of phosphoric acid monomer and diethyl methacrylate. It can be used as a chelating absorbent due to its high affinity towards metal ions.Phosphoric acid 2-hydroxyethyl methacrylate ester is used in surface functionalization of polytetrafluoroethylene (PTFE) for craniofacial applications.

Specifications

CAS 52628-03-2
Density 1.37 g/mL (at 25°C)
Linear Formula C6H10O3 -+ xH3PO4
Refractive Index n20/D 1.4688
Quantity 250 mL
Percent Purity 90%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.