Learn More
Hach Company Phosphoric Acid, 85%, 2.5L
Catalog No. 501994444 Shop All Hach Company Products
Encompass
H3PO4, Phosphoric Acid ACS Grade, CAS No. 7664-38-2, FW = 98.00. Assay: ≥85.0% 2.5 L Bottle
Catalog No. 50-199-4444 Supplier Hach Company Supplier No. 76215
H3PO4, Phosphoric Acid ACS Grade, CAS No. 7664-38-2, FW = 98.00. Assay: ≥85.0% 2.5 L Bottle