Learn More
Medchemexpress LLC Phosphoric acid, butyl diphenyl ester | 2752-95-6 | MFCD00411621 | >95.0% | 306.29 g/mol | C16H19O4P | 10 MG
SDP
Catalog No. 5000284478
Encompass_Preferred
Butyl diphenyl phosphate is an organophosphate used primarily as a flame retardant and plastic additive in materials research and formulation studies. It is provided as a research reagent and is identified by CAS 2752-95-6, molecular formula C16H19O4P, and molecular weight 306.29 g/mol.
  • Organophosphate flame retardant for polymer formulations.
  • Used as a plastic additive to improve fire resistance.
  • Well-defined chemical identifiers (CAS 2752-95-6, C16H19O4P).
  • Typical purity greater than 95% (HPLC) from commercial suppliers.
  • Available in small research quantities suitable for analytical work.
Catalog No. 50-002-84478 Supplier Medchemexpress LLC Supplier No. HYW54922710MG
Butyl diphenyl phosphate is an organophosphate used primarily as a flame retardant and plastic additive in materials research and formulation studies. It is provided as a research reagent and is identified by CAS 2752-95-6, molecular formula C16H19O4P, and molecular weight 306.29 g/mol.
  • Organophosphate flame retardant for polymer formulations.
  • Used as a plastic additive to improve fire resistance.
  • Well-defined chemical identifiers (CAS 2752-95-6, C16H19O4P).
  • Typical purity greater than 95% (HPLC) from commercial suppliers.
  • Available in small research quantities suitable for analytical work.