Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphoric acid, diethyl ester | 598-02-7 | MFCD00039889 | 98.1% | 154.10 g/mol | C4H11O4P | 100 MG
SDP

Catalog No. 5000245731
Encompass_Preferred

Diethyl phosphate is an organophosphorus metabolite commonly used as an analytical reference standard for biochemical and toxicological research. It is supplied as a high-purity liquid suitable for HPLC analysis and assay verification, with recommended storage conditions for long-term stability.

  • Purity 98.1% (HPLC).
  • CAS 598-02-7.
  • Molecular formula C4H11O4P; molecular weight 154.10 g/mol.
  • Appearance colorless to light yellow liquid.
  • Suitable as an analytical standard and metabolite reference for toxicology and biochemical studies.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size 100 MG.

Catalog No. 50-002-45731 Supplier Medchemexpress LLC Supplier No. HY101417100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl phosphate is an organophosphorus metabolite commonly used as an analytical reference standard for biochemical and toxicological research. It is supplied as a high-purity liquid suitable for HPLC analysis and assay verification, with recommended storage conditions for long-term stability.

  • Purity 98.1% (HPLC).
  • CAS 598-02-7.
  • Molecular formula C4H11O4P; molecular weight 154.10 g/mol.
  • Appearance colorless to light yellow liquid.
  • Suitable as an analytical standard and metabolite reference for toxicology and biochemical studies.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.