Learn More
Medchemexpress LLC Phosphoric acid, dipotassium salt, trihydrate | 16788-57-1 | MFCD00149926 | 99.2% | 228.22 g/mol | H7K2O7P | 500 G
SDP
Catalog No. 5000208386
Encompass_Preferred
Dipotassium hydrogen phosphate trihydrate is an inorganic salt commonly used as a buffering agent and source of potassium and phosphate in laboratory, pharmaceutical, and industrial applications. It is supplied as a white to off-white crystalline solid and is used in buffer preparation, cell culture media formulation, and analytical reagent systems.
  • Used as a buffer and pH regulator in aqueous systems.
  • Provides a source of phosphate and potassium ions.
  • Suitable for buffer preparation and cell culture media formulation.
  • Off-white crystalline solid with high purity.
  • Available in laboratory pack sizes including 500 g.
Catalog No. 50-002-08386 Supplier Medchemexpress LLC Supplier No. HYW088069500G
Dipotassium hydrogen phosphate trihydrate is an inorganic salt commonly used as a buffering agent and source of potassium and phosphate in laboratory, pharmaceutical, and industrial applications. It is supplied as a white to off-white crystalline solid and is used in buffer preparation, cell culture media formulation, and analytical reagent systems.
  • Used as a buffer and pH regulator in aqueous systems.
  • Provides a source of phosphate and potassium ions.
  • Suitable for buffer preparation and cell culture media formulation.
  • Off-white crystalline solid with high purity.
  • Available in laboratory pack sizes including 500 g.