Learn More
Medchemexpress LLC Phosphoric acid tributyl ester-d27 | 61196-26-7 | 100.0% | 293.48 g/mol | C12D27O4P | 1 MG
SDP
Catalog No. 5000226728
Encompass_Preferred
Tributyl phosphate-d27 is the deuterium-labeled analogue of tributyl phosphate used as an internal standard and tracer for quantitative analysis in NMR, GC-MS, and LC-MS. It is supplied as a high-purity stable isotope reference for analytical and research applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • High purity (≈99.95%).
  • Chemical formula C12D27O4P; molecular weight 293.48 g/mol.
  • Available in mg-scale quantities for reference use.
  • Supplied with a certificate of analysis for batch verification.
Catalog No. 50-002-26728 Supplier Medchemexpress LLC Supplier No. HY117600S1MG
Tributyl phosphate-d27 is the deuterium-labeled analogue of tributyl phosphate used as an internal standard and tracer for quantitative analysis in NMR, GC-MS, and LC-MS. It is supplied as a high-purity stable isotope reference for analytical and research applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • High purity (≈99.95%).
  • Chemical formula C12D27O4P; molecular weight 293.48 g/mol.
  • Available in mg-scale quantities for reference use.
  • Supplied with a certificate of analysis for batch verification.