Learn More
Medchemexpress LLC Phosphoric acid tributyl ester-d27 | 61196-26-7 | 100.0% | 293.48 g/mol | C12D27O4P | 10 MG
SDP
Catalog No. 5000226768
Encompass_Preferred
Tributyl phosphate-d27 is the deuterium-labeled form of tributyl phosphate used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is provided as a high-purity, isotopically labeled reference material for analytical and research applications.
  • High purity (99.95%) and defined isotopic labeling for accurate quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS analyses.
  • Soluble in DMSO at ≥ 100 mg/mL for convenient sample preparation.
  • Examples of in vivo formulations achieve ≥ 2.5 mg/mL as clear solutions.
  • Stable under recommended storage conditions: -20°C (pure) and -80°C (in solvent) for specified durations.
Catalog No. 50-002-26768 Supplier Medchemexpress LLC Supplier No. HY117600S10MG
Tributyl phosphate-d27 is the deuterium-labeled form of tributyl phosphate used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is provided as a high-purity, isotopically labeled reference material for analytical and research applications.
  • High purity (99.95%) and defined isotopic labeling for accurate quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS analyses.
  • Soluble in DMSO at ≥ 100 mg/mL for convenient sample preparation.
  • Examples of in vivo formulations achieve ≥ 2.5 mg/mL as clear solutions.
  • Stable under recommended storage conditions: -20°C (pure) and -80°C (in solvent) for specified durations.