Learn More
Cayman Chemical PhosphorScent Oxygen Probe 1ea
SDP
Catalog No. 502893078
Encompass_Preferred
A TTR kinetic stabilizer selectively binds to wild-type TTR and TTRV122I with negative cooperativity (Kd1s 4.8 and 6.2 nM respectively Kd2s 314 and 139 nM respectively) over COX-1 COX-2 and the thyroid hormone nuclear receptor (IC50s >10 UM for all) inhibits pH-induced amyloidogenesis of wild-type TTR and TTRV122I indicating tetramer stabilization stabilizes TTRV122I in serum isolated from patients with familial amyloid cardiomyopathy at 10 UM
Catalog No. 50-289-3078 Supplier Cayman Chemical Supplier No. 6008011 EA
A TTR kinetic stabilizer selectively binds to wild-type TTR and TTRV122I with negative cooperativity (Kd1s 4.8 and 6.2 nM respectively Kd2s 314 and 139 nM respectively) over COX-1 COX-2 and the thyroid hormone nuclear receptor (IC50s >10 UM for all) inhibits pH-induced amyloidogenesis of wild-type TTR and TTRV122I indicating tetramer stabilization stabilizes TTRV122I in serum isolated from patients with familial amyloid cardiomyopathy at 10 UM