Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 10mCi (370MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 502809332
Encompass
Phosphorus-32 Radionuclide, 10mCi (370MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 50-280-9332 Supplier Revvity Health Sciences Inc Supplier No. NEX053010MC
Phosphorus-32 Radionuclide, 10mCi (370MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water