Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 25mCi (925MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water, Concentration: 10mCi/mL
Catalog No. 502810484
Encompass
Phosphorus-32 Radionuclide, 25mCi (925MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water, Concentration: 10mCi/mL
Catalog No. 50-281-0484 Supplier Revvity Health Sciences Inc Supplier No. NEX053H025MC
Phosphorus-32 Radionuclide, 25mCi (925MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water, Concentration: 10mCi/mL