Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 2mCi (74MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water
Catalog No. 502810993
Encompass
Phosphorus-32 Radionuclide, 2mCi (74MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water
Catalog No. 50-281-0993 Supplier Revvity Health Sciences Inc Supplier No. NEX053H002MC
Phosphorus-32 Radionuclide, 2mCi (74MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water