Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 2mCi (74MBq), Specific Activity: 900-1100mCi (33.3-40.7GBq)/mmol, Monopotassium Phosphate In 1mL Water
Catalog No. 502810646
Encompass
Phosphorus-32 Radionuclide, 2mCi (74MBq), Specific Activity: 900-1100mCi (33.3-40.7GBq)/mmol, Monopotassium Phosphate In 1mL Water
Catalog No. 50-281-0646 Supplier Revvity Health Sciences Inc Supplier No. NEX060002MC
Phosphorus-32 Radionuclide, 2mCi (74MBq), Specific Activity: 900-1100mCi (33.3-40.7GBq)/mmol, Monopotassium Phosphate In 1mL Water