Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 50mCi (1.85GBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 502812220
Encompass
Phosphorus-32 Radionuclide, 50mCi (1.85GBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 50-281-2220 Supplier Revvity Health Sciences Inc Supplier No. NEX053050MC
Phosphorus-32 Radionuclide, 50mCi (1.85GBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water