Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water, Concentration: 10mCi/mL
Supplier:  Revvity Health Sciences Inc NEX053H005MC
Encompass
Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in Water, Concentration: 10mCi/mL
Catalog No. 50-281-1008