Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 900-1100mCi (33.3-40.7GBq)/mmol, Monopotassium Phosphate In 1mL Water
Supplier:  Revvity Health Sciences Inc NEX060005MC
Encompass
Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 900-1100mCi (33.3-40.7GBq)/mmol, Monopotassium Phosphate In 1mL Water
Catalog No. 50-281-2317