Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 502810963
Encompass
Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 50-281-0963 Supplier Revvity Health Sciences Inc Supplier No. NEX053005MC
Phosphorus-32 Radionuclide, 5mCi (185MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water