Learn More
Strem, An Ascensus Company 500mg. Di-t-butyl(n-hexyl)phosphonium tetrafluoroborate, 98%. MFCD24842681.
SDP
Catalog No. NC1868638
Encompass
500mg. Di-t-butyl(n-hexyl)phosphonium tetrafluoroborate, 98%. MFCD24842681. Molecular Weight: 318.18. Molecular Formula: C14H32BF4P. Color/form: white pwdr. Strem# 15-7230. http://www.strem.com/catalog/v/15-7230/
Catalog No. NC1868638 Supplier Strem, An Ascensus Company Supplier No. 157230500MG
500mg. Di-t-butyl(n-hexyl)phosphonium tetrafluoroborate, 98%. MFCD24842681. Molecular Weight: 318.18. Molecular Formula: C14H32BF4P. Color/form: white pwdr. Strem# 15-7230. http://www.strem.com/catalog/v/15-7230/