Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus ICP Standard, 1000 ppm P in H2O, Ricca Chemical
SDP

Catalog No. PP1KW100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PP1KW-100 100 mL Natural LDPE Bottle
PP1KW-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PP1KW-100 Supplier Ricca Chemical Company Supplier No. PP1KW100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-76-1 , 7732-18-5
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 98.0, 0.36
CAS Max % 100.0, 0.38
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Type Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg P (1, 000ppm P)NH4H2PO4 in H2O
Chemical Name or Material Phosphorus ICP Standard
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.