Learn More
Strem, An Ascensus Company CAS# 7789-60-8. 100g. Phosphorus(III) bromide, 97+%. MFCD00011436
SDP
Catalog No. 5090114222
Encompass
CAS# 7789-60-8. 100g. Phosphorus(III) bromide, 97+%. MFCD00011436. Molecular Weight: 270.70. Molecular Formula: PBr3. Color/form: colorless to pale yellow liq. Strem# 93-1550. http://www.strem.com/catalog/v/93-1550/
Catalog No. 50-901-14222 Supplier Strem, An Ascensus Company Supplier No. 931550100G
CAS# 7789-60-8. 100g. Phosphorus(III) bromide, 97+%. MFCD00011436. Molecular Weight: 270.70. Molecular Formula: PBr3. Color/form: colorless to pale yellow liq. Strem# 93-1550. http://www.strem.com/catalog/v/93-1550/