Phosphorus(III) chloride, 98% min., Alfa Aesar™

Manufacturer: Alfa Aesar 4443618

Quantity 50g

 View more versions of this product

Catalog No. AA4443618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7719-12-2
    Physical Form Liquid
    Assay Percent Range 98% min.
    Molecular Formula PCl3
    Formula Weight 137.33
    MDL Number MFCD00011438
    Chemical Name or Material Phosphorus(III) chloride

    Specifications

    Physical Form Liquid
    Formula Weight 137.33
    Quantity 50g
    Color Yellow
    DOT Information DOT Class: 6.1, Packaging Group: I
    Health Hazard 2 Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    EINECS Number 231-749-3
    Molecular Formula PCl3
    CAS 7719-12-2
    Odor Acrid
    Assay Percent Range 98% min.
    MDL Number MFCD00011438
    Vapor Pressure 95mm Hg at 20°C
    Health Hazard 1 H300-H330-H314-H318-H373: Fatal if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Phosphorus(III) chloride

    Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H300-H330-H314-H318-H373: Fatal if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. May cause damage to organs through prolonged or repeated exposure.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit