Learn More
Strem, An Ascensus Company CAS# 1810068-31-5. 250mg. 2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]-2',6'-di-i-propoxy-1,1'-biphenyl, 98%.
SDP
Supplier:  Strem, An Ascensus Company 153020250MG
Encompass
CAS# 1810068-31-5. 250mg. 2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]-2',6'-di-i-propoxy-1,1'-biphenyl, 98%. . Molecular Weight: 786.58. Molecular Formula: C36H31F12O4P. Color/form: white to off-white pwdr. Strem# 15-3020. http://www.strem.com/catalog/v/15-3020/
Catalog No. 50-204-5250