Learn More
Strem, An Ascensus Company CAS# 1810068-30-4. 500mg. 2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]-2',6'-dimethylamino-1,1'-biphenyl, 98%.
SDP
Supplier:  Strem, An Ascensus Company 153015500MG
Encompass
CAS# 1810068-30-4. 500mg. 2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]-2',6'-dimethylamino-1,1'-biphenyl, 98%. . Molecular Weight: 756.56. Molecular Formula: C34H29F12N2O2P. Color/form: white to off-white pwdr. Strem# 15-3015. http://www.strem.com/catalog/v/15-3015/
Catalog No. 50-204-5248