Learn More
Strem, An Ascensus Company CAS 192138-05-9. 500mg. (R)-()-22-Bis[di(35-di-t-butylphenyl)phosphino]-66-dimethoxy-11-biphenyl min. 97%. MFCD09753001
SDP
Supplier:  Strem, An Ascensus Company 150044500MG
Encompass
CAS 192138-05-9. 500mg. (R)-()-22-Bis[di(35-di-t-butylphenyl)phosphino]-66-dimethoxy-11-biphenyl min. 97%. MFCD09753001. Molecular Weight 1031.49. Molecular Formula C70H96O2P2. Color/form white pwdr. Strem 15-0044. http//www.strem.com/catalog/v/15-0044/
Catalog No. 50-211-0719