Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphorus pentachloride r1KG
Catalog No. 502898987
Encompass
Phosphorus pentachloride is a light yellow colored phosphorous halide. It participates in the synthesis of thiazoline analogs.
Catalog No. 50-289-8987 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1577751KG
Phosphorus pentachloride is a light yellow colored phosphorous halide. It participates in the synthesis of thiazoline analogs.