Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus pentasulfide, 98+%, Thermo Scientific Chemicals

Catalog No. p-3213051
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196725000 500 g Glass bottle
AC196720010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196725000 Supplier Thermo Scientific Chemicals Supplier No. 196725000
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-80-3
Molecular Formula P4S10
Molecular Weight (g/mol) 444.48
MDL Number MFCD00011441
InChI Key CYQAYERJWZKYML-UHFFFAOYSA-N
Synonym phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech
PubChem CID 14817
SMILES P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3

Specifications

Boiling Point 514.0°C
Melting Point 282.0°C to 286.0°C
Color Yellow to Green
Physical Form Powder
Quantity 500 g
Assay Percent Range 98% min
Linear Formula P4S10
Flash Point >100°C
Fieser 01,870; 03,226; 04,389; 05,534; 06,470; 07,290; 08,401; 09,374; 10,320
Merck Index 15,7466
Solubility Information Solubility in water: reacts forming h3po4 and h2s. Other solubilities: soluble in solutions of alkali hydroxides, soluble in carbon disulfide, reacts with alcohols and acids
Formula Weight 444.48
Percent Purity ≥98%
Packaging Glass bottle
Density 2.08
Chemical Name or Material Phosphorus pentasulfide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.
Contact with water liberates toxic
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 215-242-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.