Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Phosphorus Standard, 50 ppm P (153 ppm PO43-), Ricca Chemical
SDP

Catalog No. R58300001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R58300001A 1 L
5830-4 120 mL
5830-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R58300001A Supplier Ricca Chemical Company Supplier No. 583032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Natural Poly Bottle
CAS 7778-77-0, 7732-18-5
Odor Odorless
Assay Percent Range 50.00 ± 0.25ppm
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Boiling Point 100°C
Solubility Infinitely soluble in water
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
Solvent or Matrix Water
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
Quantity 1 L
PubChem CID 516951
ChEBI CHEBI:63036
Chemical Name or Material Phosphate Standard, 50ppm P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.