Learn More
SPEX CERTIPREP LLC 3P PHOSPHORUS STD 1000PPM 100ML
Catalog No. NC9423618
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Phosphorus in H2O; Single component Ion Chromatography Standard, 18 month expiry date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9423618 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPM100
(HPS by Zeptometrix) 1000 ug/mL Phosphorus in H2O; Single component Ion Chromatography Standard, 18 month expiry date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.