Learn More
Hach Company Phosphorus (Reactive and Total) TNTplus Vial Test, UHR (6-60 mg/L PO₄), 25 Tests
Catalog No. 501998137 Shop All Hach Company Products
Encompass
Phosphorus TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Reactive (ortho) and Total Phosphorus (phosphate) by the Ascorbic Acid method.
Catalog No. 50-199-8137 Supplier Hach Company Supplier No. TNT845
Phosphorus TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Reactive (ortho) and Total Phosphorus (phosphate) by the Ascorbic Acid method.