Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus (total), Standard Solution 7, CRM, MilliporeSigma™

Catalog No. M1250490100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1250490100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1250490100 Supplier MilliporeSigma Supplier No. 1250490100
Add to Cart
Add to Cart

Specifications

Packaging Plastic Bottle
Physical Form Aqueous solution
Concentration 75 mg/L PO4-P
Color Colorless
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 75mg/L PO4-P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.