Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphorus trichloride

Catalog No. 310115100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-011-5100G 100 g
31-011-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-011-5100G Supplier Sigma Aldrich Supplier No. 310115100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphorus trichloride

Specifications

Melting Point -112°C
Density 1.574 g/mL (at 20°C)
Boiling Point 74°C to 78°C
Molecular Formula Cl3P
Refractive Index n20/D 1.5148 (literature)
Linear Formula PCl3
Quantity 100 g
MDL Number MFCD00011438
Synonym Phosphorus(III) chloride
Molecular Weight (g/mol) 137.33
Percent Purity 99.999% trace metals basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.