Learn More
Strem, An Ascensus Company CAS# 10026-13-8. 250G. Phosphorus(V) chloride, 98%. MFCD00011439
SDP
Catalog No. 501963613
Encompass
CAS# 10026-13-8. 250G. Phosphorus(V) chloride, 98%. MFCD00011439. Molecular Formula: PCl5. Molecular Weight: 208.24. Color/Form: white to pale yellow xtl. Stability: moisture sensitive. Strem# 93-1545. www.strem.com/catalog/v3/93-1545/
Catalog No. 50-196-3613 Supplier Strem, An Ascensus Company Supplier No. 931545250G
CAS# 10026-13-8. 250G. Phosphorus(V) chloride, 98%. MFCD00011439. Molecular Formula: PCl5. Molecular Weight: 208.24. Color/Form: white to pale yellow xtl. Stability: moisture sensitive. Strem# 93-1545. www.strem.com/catalog/v3/93-1545/