Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals

Catalog No. p-3257053
Change view
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1052322 100 g
AA1052336 500 g
AA1052357 4 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1052322 Supplier Thermo Scientific Chemicals Supplier No. 01052322
Only null left
Add to Cart

For Use As Dehydrating and Chlorinating Agent, Catalyst

Phosphorus(V) chloride can be used for both oxidative and substitutive chlorinations, & for both organic and inorganic compounds. Dehydrating agent. Precursor to Vilsmeier reagent, lithium hexafluorophosphate electrolyite for lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-13-8
Molecular Formula Cl5P
Molecular Weight (g/mol) 208.224
MDL Number MFCD00011439
InChI Key UHZYTMXLRWXGPK-UHFFFAOYSA-N
Synonym phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
PubChem CID 24819
ChEBI CHEBI:30335
IUPAC Name pentachloro-$l^{5}-phosphane
SMILES P(Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 162° (sublimation)
Color Yellow
Physical Form Solid
Quantity 100 g
Assay Percent Range 98%
Solubility Information Soluble in carbon disulfide and carbon tetrachloride.
Formula Weight 208.24
Odor Pungent
Sensitivity Moisture sensitive
Appearance Yellowish crystals
Vapor Pressure 0.02 hPa at 20°C
Density 2.114 g/cm3 at 20°C
Chemical Name or Material Phosphorus(V) chloride
Show More Show Less
Hazard Category H302-H314-H330-H373
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 233-060-3
RTECSNumber TB6125000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.