Learn More
Strem, An Ascensus Company CAS# 1314-56-3. 500g. Phosphorus(V) oxide, 98+% (ACS). MFCD00011440
SDP
Catalog No. 5090114220
Encompass
CAS# 1314-56-3. 500g. Phosphorus(V) oxide, 98+% (ACS). MFCD00011440. Molecular Weight: 141.95. Molecular Formula: P2O5. Color/form: white pwdr. Strem# 93-1548. http://www.strem.com/catalog/v/93-1548/
Catalog No. 50-901-14220 Supplier Strem, An Ascensus Company Supplier No. 931548500G
CAS# 1314-56-3. 500g. Phosphorus(V) oxide, 98+% (ACS). MFCD00011440. Molecular Weight: 141.95. Molecular Formula: P2O5. Color/form: white pwdr. Strem# 93-1548. http://www.strem.com/catalog/v/93-1548/