Learn More
Strem, An Ascensus Company CAS# 1799401-53-8. 5g. 1,3-Bis(dicyclopentylphosphonium)propane bis(tetrafluoroborate), min. 97%. MFCD22666033
SDP
Catalog No. NC1866484
Encompass
CAS# 1799401-53-8. 5g. 1,3-Bis(dicyclopentylphosphonium)propane bis(tetrafluoroborate), min. 97%. MFCD22666033. Molecular Weight: 556.15. Molecular Formula: (C5H9)2PH+CH2CH2CH2PH+(C5H9)2 (BF4)2. Color/form: white pwdr. Strem# 15-9572. http://www.strem.com/catalog/v/15-9572/
Catalog No. NC1866484 Supplier Strem, An Ascensus Company Supplier No. 1595725G
CAS# 1799401-53-8. 5g. 1,3-Bis(dicyclopentylphosphonium)propane bis(tetrafluoroborate), min. 97%. MFCD22666033. Molecular Weight: 556.15. Molecular Formula: (C5H9)2PH+CH2CH2CH2PH+(C5H9)2 (BF4)2. Color/form: white pwdr. Strem# 15-9572. http://www.strem.com/catalog/v/15-9572/