Learn More
Meso Scale Discovery PhosphoS6RP Ser240/244 WCL20
SDP
Catalog No. 5000079228
Encompass
The Meso Scale Discovery Phospho-S6RP (Ser240/244) Whole Cell Lysate Kit measures phosphorylated S6 Ribosomal Protein (Ser240/244) in cell lysates via electrochemiluminescent detection It includes a 96-well plate with capture and detection antibodies buffers blocking solution and controls Detecting as little as 78 pg/mL it s ideal for analyzing S6RP phosphorylation in response to PI3K/Akt/mTOR pathway activators The kit provides all necessary reagents for high-throughput quantification from cell lysates
Catalog No. 50-000-79228 Supplier Meso Scale Discovery Supplier No. K150DGD3
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho-S6RP (Ser240/244) Whole Cell Lysate Kit measures phosphorylated S6 Ribosomal Protein (Ser240/244) in cell lysates via electrochemiluminescent detection It includes a 96-well plate with capture and detection antibodies buffers blocking solution and controls Detecting as little as 78 pg/mL it s ideal for analyzing S6RP phosphorylation in response to PI3K/Akt/mTOR pathway activators The kit provides all necessary reagents for high-throughput quantification from cell lysates