Learn More
Enzo Life Sciences Phosphoserine monoclonal antibody (1C8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804162C100
Encompass_Preferred
Clone: 1C8. Isotype: IgM. Immunogen: Phosphoserine containing peptides conjugated to KLH. Species reactivity: Species independent. Specificity: Recognizes a broad range of serine-phosphorylated proteins in crude cell extracts, interacting with phosphoserine surrounded by positive or neutral aa (e.g. substrates of PKA, PKB, PKC, PKG etc.). Please note that phosphoserine detection by MAbs always depends on the surrounding aa sequence. Applications: ELISA, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9807