Learn More
Enzo Life Sciences Phosphoserine polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPST210E
Encompass_Preferred
Host: Rabbit. Immunogen: Phosphoserine conjugated to KLH. Species reactivity: Species independent. Applications: ELISA, IHC, IP, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS with 50% glycerol.
Catalog No. 50-200-7299