Learn More
Meso Scale Discovery PhosphoThr231/Tau TTL Kit
SDP
Catalog No. 5000078228
Encompass
The Meso Scale Discovery Phospho-Thr231/Total Tau Kit measures phosphorylated Thr231 and total Tau in samples like CSF serum plasma cell lysates and brain homogenates It includes a 96-well plate with capture antibodies for phosphorylated Thr231 Tau and total Tau using electroluminescence for high sensitivity The kit contains a plate detection antibodies read buffer calibrator and sample diluent The assay runs in under 4 hours enabling quantitative measurement of Tau phosphorylation at Thr231 linked to neurofibrillary tangle pathology in Alzheimer s disease
Catalog No. 50-000-78228 Supplier Meso Scale Discovery Supplier No. K15121D1
Add to Cart
Add to Cart
The Meso Scale Discovery Phospho-Thr231/Total Tau Kit measures phosphorylated Thr231 and total Tau in samples like CSF serum plasma cell lysates and brain homogenates It includes a 96-well plate with capture antibodies for phosphorylated Thr231 Tau and total Tau using electroluminescence for high sensitivity The kit contains a plate detection antibodies read buffer calibrator and sample diluent The assay runs in under 4 hours enabling quantitative measurement of Tau phosphorylation at Thr231 linked to neurofibrillary tangle pathology in Alzheimer s disease