Learn More
Meso Scale Discovery PhosphoThr231/Tau TTL Kit
SDP
Catalog No. 5000078229
Encompass
The Meso Scale Discovery Phospho-Thr231/Total Tau Kit is a multiplex immunoassay for measuring phosphorylated Thr231 tau and total tau in samples like CSF serum plasma cell lysates and brain homogenates It features a 96-well plate with capture antibodies and uses MSD s electrochemiluminescence for sensitive quantification Ideal for Alzheimer s research it supports high-throughput analysis of tau biomarkers includes all necessary reagents and processes up to 80 samples in duplicate
This item is not returnable. View return policy
Catalog No. 50-000-78229 Supplier Meso Scale Discovery Supplier No. K15121D2
The Meso Scale Discovery Phospho-Thr231/Total Tau Kit is a multiplex immunoassay for measuring phosphorylated Thr231 tau and total tau in samples like CSF serum plasma cell lysates and brain homogenates It features a 96-well plate with capture antibodies and uses MSD s electrochemiluminescence for sensitive quantification Ideal for Alzheimer s research it supports high-throughput analysis of tau biomarkers includes all necessary reagents and processes up to 80 samples in duplicate