Learn More
Meso Scale Discovery PhosphoThr231/Tau TTL20 Plate
SDP
Catalog No. 5000082419
Encompass
The Meso Scale Discovery Phospho-Thr231/Total Tau Kit is a multiplex immunoassay kit that allows for the simultaneous measurement of phosphorylated Thr231 tau and total tau in biological samples such as CSF serum plasma cell lysates and brain homogenates The kit contains a 96-well plate coated with capture antibodies specific for phosphorylated Thr231 tau and total tau It utilizes MSD s electrochemiluminescence detection technology to provide sensitive and accurate quantification of both analytes from a single small sample volume The kit includes all necessary reagents and can process up to 80 samples in duplicate It is suitable for studying Alzheimer s disease and other tauopathies
Catalog No. 50-000-82419 Supplier Meso Scale Discovery Supplier No. K15121D3
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho-Thr231/Total Tau Kit is a multiplex immunoassay kit that allows for the simultaneous measurement of phosphorylated Thr231 tau and total tau in biological samples such as CSF serum plasma cell lysates and brain homogenates The kit contains a 96-well plate coated with capture antibodies specific for phosphorylated Thr231 tau and total tau It utilizes MSD s electrochemiluminescence detection technology to provide sensitive and accurate quantification of both analytes from a single small sample volume The kit includes all necessary reagents and can process up to 80 samples in duplicate It is suitable for studying Alzheimer s disease and other tauopathies