Learn More
ECM Bioscience Anti-Phosphothreonine Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP4641
Encompass_Preferred
This antibody was cross-adsorbed to unphosphorylated peptide then affinity purified using phospho-threonine peptide (without carrier). The antibody detects many serine or threonine phosphorylated proteins by western blot and ELISA. The immunogen was Phosphothreonine synthetic peptides coupled to carrier protein. Accession #. Applications = WB, E, IP, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = phosphotyrosine, phoshoserine, phoshothreonine
Catalog No. 50-172-0853