Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotungstic Acid AR Crystal, Macron Fine Chemicals™

Catalog No. MK2824500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
30 g
125 g
500 g
Packaging:
POLYSTORMOR
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-2824-500 500 g POLYSTORMOR
MK-2824-30 30 g POLYSTORMOR
MK-2824-125 125 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MK-2824-500 Supplier Avantor Supplier No. 282404
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 12067-99-1
Molecular Formula P2O5·24WO3·nH2O

Specifications

Packaging POLYSTORMOR
Quantity 500 g
Physical Form Crystalline
Chemical Name or Material Phosphotungstic Acid, n-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.