Learn More
Electron Microscopy Sciences PTA (Phosphotungstic Acid), Crystal 100 GR
SDP
Catalog No. 5033223
Encompass_Preferred
(Tungstophosphoric Acid)
CAS #12501-23-4 H₃[P(W₃O₁₀)₄] F.W. 2880.17 aq
Tungstophosphoric Acid is used as a fixative and widely as a Negative Stain. References
Issidorides, M.R., and Kasorchis, T. (1981). Dispersed and compact chromatin demonstrated with a new EM Method: phosphotungstic acid-hematoxylin block-staining. Histochemistry 73,21.
Catalog No. 50-332-23 Supplier Electron Microscopy Sciences Supplier No. 19500
(Tungstophosphoric Acid)
CAS #12501-23-4 H₃[P(W₃O₁₀)₄] F.W. 2880.17 aq
Tungstophosphoric Acid is used as a fixative and widely as a Negative Stain. References
Issidorides, M.R., and Kasorchis, T. (1981). Dispersed and compact chromatin demonstrated with a new EM Method: phosphotungstic acid-hematoxylin block-staining. Histochemistry 73,21.