Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphotungstic acid hydrate

Catalog No. 45597010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
10 g
50 g

Catalog No. 45-597-010G

Supplier: Sigma Aldrich 45597010G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphotungstic acid hydrate

Specifications

CAS 12501-23-4
Melting Point ∼95°C
Molecular Formula H3O40PW12 -+ xH2O
Linear Formula H3[P(W3O10)4 ] -+ xH2O
Quantity 10 g
MDL Number MFCD00149915
Synonym Tungstophosphoric acid
Molecular Weight (g/mol) 2880.05
Percent Purity 99.995% trace metals basis (Purity excludes up to 300 ppm Si)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.